N,N-Diethyl-m-toluamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11457-0025
11457-1000
11457-5000
TS11457-0025EA
211
USD
TS11457-0025
TS11457-1000
TS11457-5000
N,N-Diethyl-m-toluamide 98%
N,N-Diethyl-m-toluamide
Formula:
C₁₂H₁₇NO Boiling Pt: 111 °C (1 mmHg) Density: 0.9955 g/cm³ Flash Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009046 CAS Number: 134-62-3 EINECS: 205-149-7 Merck Index: 13,02876 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5240 (25°C, 589 nm) |
Water | ≤0.3 % |
GC | ≥97.5 % |
Color scale | ≤100 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...