Diethyl phosphonate 98%

Supplier: Thermo Scientific Chemicals
Danger

11450-2500
TS11450-2500EA 53 USD
TS11450-2500
Diethyl phosphonate 98%
Diethyl phosphonate
Formula: C₄H₁₁O₃P
Boiling Pt: 60 °C (6mbar)
Melting Pt: –70 °C
Density: 1.08 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00044573
CAS Number: 762-04-9
EINECS: 212-091-6

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4090 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR