Diethyl malonate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester

11445-0010 11445-0025 11445-0100 11445-0250 11445-5000
TS11445-0010EA 69 USD
TS11445-0010 TS11445-0025 TS11445-0100 TS11445-0250 TS11445-5000
Diethyl malonate 99+%
Diethyl malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199
Melting Pt: -50
Density: 1.055 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20 °C, 589 nm)
Density (20°C) 1.052 to 1.058 g/ml
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR