Silver diethyldithiocarbamate 99% for analysis

Supplier: Thermo Scientific Chemicals
Warning

11429-1000 AC11429-0050 AC11429-0250
TS11429-1000EA 814 USD
TS11429-1000 TS11429-0050 TS11429-0250
Silver diethyldithiocarbamate 99% for analysis
Silver diethyldithiocarbamate
Formula: (C₂H₅)₂NCSSAg
Melting Pt: 172…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7

Order Now

Specification Test Results

Appearance Green to khaki Powder
Infrared spectrum Conforms
Solubility (0.5% in pyridine) clear
Titration with NH4SCN ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR