Diethyl aminomalonate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2-aminomalonate hydrochloride

11404-0250 11404-1000
TS11404-0250EA 60 USD
TS11404-0250 TS11404-1000
Diethyl aminomalonate hydrochloride 98%
Diethyl aminomalonate hydrochloride
Formula: C₇H₁₄ClNO₄
MW: 211.65 g/mol
Melting Pt: 166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012510
CAS Number: 13433-00-6
EINECS: 236-556-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Titration with NaOH ≥99.0 %
Chloride (Cl) ≤10 ppm
Oxalate (C2O4) (limit about 0.1%) passes test
Phosphate (PO4) ≤10 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.002 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR