Cyclohexanone 99.8%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

11119-0010 111190025 111190250 111190251
TS11119-0010EA 69 USD
TS11119-0010 TS111190025 TS111190250 TS111190251
Cyclohexanone 99.8%, Extra Pure
Cyclohexanone
Formula: C₆H₁₀(=O)
Boiling Pt: 155 °C (760 mmHg)
Melting Pt: –47 °C
Density: 0.947 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4520 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥99.75 %
Color scale ≤20 APHA
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.005 %
Manganese (Mn) ≤0.002 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.05 %
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR