1,2-Cyclohexanediol cis- and trans- mixture 98%

Synonyms: Cyclohexane-1,2-diol

11113-0250
TS11113-0250EA 108 USD
TS11113-0250 TS11113-1000
1,2-Cyclohexanediol cis- and trans- mixture 98%
1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 237
Melting Pt: 73
Flash Pt: >110 °C
MDL Number: MFCD00003861
CAS Number: 931-17-9
EINECS: 213-229-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 77 °C
GC ≥97.5 %
Absorption Ratio P-15/P+15 1 to 1.12
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR