4-Chlorobutyryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

10890-0010 10890-2500
TS10890-0010EA 118 USD
TS10890-0010 TS10890-2500
4-Chlorobutyryl chloride 98%
4-Chlorobutyryl chloride
Formula: C₄H₆Cl₂O
Boiling Pt: 173…174 °C (760 mmHg)
Melting Pt: –47 °C
Density: 1.25 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000754
CAS Number: 4635-59-0
EINECS: 225-059-1
UN: 2922
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4605 to 1.4625 (20°C, 589 nm)
GC ≥99.0 %
Solubility (in water) Passes test
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.02 % 2-Butanone
Titrable acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR