3-Chlorobenzoic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

10866-0250 10866-1000 10866-5000
TS10866-0250EA 33 USD
TS10866-0250 TS10866-1000 TS10866-5000
3-Chlorobenzoic acid 99+%
3-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 281
Melting Pt: 122
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002491
CAS Number: 535-80-8
EINECS: 208-618-4
Merck Index: 13,02142

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 157 °C
HPLC ≥99.0 %
Titration with NaOH 99.0 to 100.5 %
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR