tert-Butylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

10782-0010 10782-0025 10782-0050 10782-0100
TS10782-0010EA 64 USD
TS10782-0010 TS10782-0025 TS10782-0050 TS10782-0100
tert-Butylamine 99%
tert-Butylamine
Formula: C₄H₁₁N
Boiling Pt: 46 °C (760 mmHg)
Melting Pt: –67 °C
Density: 0.69 g/cm³
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Merck Index: 13,01544

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.5 %
Color scale ≤10 APHA
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.002 meq/g
Insoluble matter ≤0.005 %
Starch Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR