4-Bromopyridinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromopyridine HCl, 4-Bromopyridine hydrochloride
AC10742-0050
AC10742-0250
TS10742-0050EA
110
USD
TS10742-0050
TS10742-0250
4-Bromopyridinium chloride 98%
4-Bromopyridinium chloride
Formula:
C₅H₅BrClN Melting Pt: 313…315 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012828 CAS Number: 19524-06-2 EINECS: 243-128-4 |
Specification Test Results
Appearance | White to light yellow-pink Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
UV | at 400 nm A: ≤0.004 |
Color scale | ≤10 APHA |
Stabilizer | Approx. 1 % w/w Ethanol |
Peroxides | ≤5 ppm (as H2O2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...