1-Bromo-2-fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromofluorobenzene
10699-0100
10699-0500
TS10699-0100EA
43
USD
TS10699-0100
TS10699-0500
1-Bromo-2-fluorobenzene 99%
1-Bromo-2-fluorobenzene
Formula:
C₆H₄BrF MW: 175 g/mol Boiling Pt: 155 Density: 1.6 g/cm³ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000282 CAS Number: 1072-85-1 EINECS: 214-018-3 UN: 1993 ADR: 3 III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5327 to 1.5347 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...