2-Bromethylamine hydrobromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide
AC10693-0010
AC10693-0025
AC10693-0250
AC10693-1000
AC10693-2500
AC10693-5000
TS10693-0010EA
806
USD
TS10693-0010
TS10693-0025
TS10693-0250
TS10693-1000
TS10693-2500
TS10693-5000
2-Bromethylamine hydrobromide 99%
2-Bromethylamine hydrobromide
Formula:
BrCH₂CH₂NH₃Br MW: 204.89 g/mol Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012886 CAS Number: 2576-47-8 EINECS: 219-924-2 |
Specification Test Results
Appearance | White to light beige Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 172 to 176 °C |
Titration Argentometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...