4'-Bromoacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
10659-0250
10659-1000
10659-5000
TS10659-0250EA
33
USD
TS10659-0250
TS10659-1000
TS10659-5000
4'-Bromoacetophenone 98%
4'-Bromoacetophenone
Formula:
C₈H₇BrO Boiling Pt: 255 °C (760 mmHg) Melting Pt: 48…52 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000105 CAS Number: 99-90-1 EINECS: 202-799-3 Merck Index: 13,01387 |
Specification Test Results
Appearance | White to light yellow Crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 48 to 52 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...