4'-Bromoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone

10659-0250 10659-1000 10659-5000
TS10659-0250EA 33 USD
TS10659-0250 TS10659-1000 TS10659-5000
4'-Bromoacetophenone 98%
4'-Bromoacetophenone
Formula: C₈H₇BrO
Boiling Pt: 255 °C (760 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
Merck Index: 13,01387

Order Now

Specification Test Results

Appearance White to light yellow Crystals or flakes
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR