2-Phenylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid

10627-0250 10627-1000
TS10627-0250EA 68 USD
TS10627-0250 TS10627-1000
2-Phenylbenzoic acid 98%
2-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
Boiling Pt: 199 °C (10 mmHg)
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002463
CAS Number: 947-84-2
EINECS: 213-432-1

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 114 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR