Benzylamine 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Aminotoluene

10585-0025 10585-0100 10585-1000 10585-5000
TS10585-0025EA 335 USD
TS10585-0025 TS10585-0100 TS10585-1000 TS10585-5000
Benzylamine 99%, pure
Benzylamine
Formula: (C₆H₅)CH₂NH₂
Boiling Pt: 182…185 °C (760 mmHg)
Melting Pt: –30 °C
Density: 0.98 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8,II
Merck Index: 13,01127

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5450 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR