Benzophenone 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
10556-0010
10556-0050
10556-5000
TS10556-0010EA
70
USD
TS10556-0010
TS10556-0050
TS10556-5000
Benzophenone 99%, pure
Benzophenone
Formula:
C₁₃H₁₀O Boiling Pt: 305 °C (760 mmHg) Melting Pt: 47…49 °C Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 Merck Index: 13,01099 |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 47 to 49 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...