Benzo[ghi]perylene 98+%

Supplier: Thermo Scientific Chemicals
Warning

10555-0050 10555-0250 10555-1000
TS10555-0050EA 43 USD
TS10555-0050 TS10555-0250 TS10555-1000
Benzo[ghi]perylene 98+%
Benzo[ghi]perylene
Formula: C₂₂H₁₂
Boiling Pt: >500 °C (760 mmHg)
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004135
CAS Number: 191-24-2
EINECS: 205-883-8

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 276 to 280 °C
GC-MS ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR