L(+)-Aspartic acid 98+%

Supplier: Thermo Scientific Chemicals

10504-0025 10504-0100 10504-1000 10504-5000
TS10504-0025EA 271 USD
TS10504-0025 TS10504-0100 TS10504-1000 TS10504-5000
L(+)-Aspartic acid 98+%
L(+)-Aspartic acid
Formula: C₄H₇NO₄
Melting Pt: >230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Sulfated ash ≤0.1 %
Specific optical rotation +24.0° to +26.0° (20°C, 589 nm) (c=8, 6 N HCl)
Titration with HClO4 ≥98.0 % (on dried substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Ninhydrin-positive substances Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR