4-Anisoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

10485-0050 10485-1000 10485-5000
TS10485-0050EA 31 USD
TS10485-0050 TS10485-1000 TS10485-5000
4-Anisoyl chloride 99%
4-Anisoyl chloride
Formula: C₈H₇ClO₂
Boiling Pt: 145 °C (14 mmHg)
Melting Pt: 22 °C
Density: 1.26 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000687
CAS Number: 100-07-2
EINECS: 202-816-4
UN: 1729
ADR: 8,II
Merck Index: 13,00676

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5800 to 1.5820 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥96.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR