4-Amino-4H-1,2,4-triazole 99%

Supplier: Thermo Scientific Chemicals
Warning

10470-0250 10470-1000
TS10470-0250EA 73 USD
TS10470-0250 TS10470-1000
4-Amino-4H-1,2,4-triazole 99%
4-Amino-4H-1,2,4-triazole
Formula: C₂H₄N₄
Melting Pt: 84 °C
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00003099
CAS Number: 584-13-4
EINECS: 209-533-5

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥98.5 %
UV at 300 nm A: ≤0.005
Residue after evaporation ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR