4-Aminopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10457-0010
10457-0050
10457-0250
10457-1000
TS10457-0010EA
817
USD
TS10457-0010
TS10457-0050
TS10457-0250
TS10457-1000
4-Aminopyridine 98%
4-Aminopyridine
Formula:
C₅H₆N₂ Boiling Pt: 273 °C (760 mmHg) Melting Pt: 157…161 °C Flash Pt: 156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 EINECS: 207-987-9 UN: 2671 ADR: 6.1,II Merck Index: 13,03964 |
Specification Test Results
Appearance | White to light yellow to beige Crystalline powder or crystals or granules |
Infrared spectrum | Conforms |
Melting point | 157 to 161 °C |
GC | ≥97.5 % |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...