4,4'-Diaminodiphenyl ether 98%

Supplier: Thermo Scientific Chemicals
Danger

10433-0250 10433-1000 10433-5000
TS10433-0050EA 27.11 USD
TS10433-0050 TS10433-0250 TS10433-1000 TS10433-5000
4,4'-Diaminodiphenyl ether 98%
4,4'-Diaminodiphenyl ether
Formula: C₁₂H₁₂N₂O
Boiling Pt: 190 °C (.1 mmHg)
Melting Pt: 189…193 °C
Flash Pt: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00007863
CAS Number: 101-80-4
EINECS: 202-977-0
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 193 °C
Titration with HClO4 ≥97.5 %
Iron (Fe) ≤500 ppm
Magnesium (Mg) ≤5000 ppm
Potassium (K) ≤500 ppm
Sodium (Na) ≤500 ppm
Strontium (Sr) ≤500 ppm
Heavy metals (ICP-OES) ≤30 ppm
Sulfur compounds (as SO4) ≤0.1 %
Insoluble matter ≤0.03 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR