2-Amino-1-phenylethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

10432-0100
TS10432-0100EA 107 USD
TS10432-0100 TS10432-0500
2-Amino-1-phenylethanol 98%
2-Amino-1-phenylethanol

Appearance: Clear yellow Viscous liquid

Formula: C₈H₁₁NO
Boiling Pt: 160 °C (17 mmHg)
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008137
CAS Number: 7568-93-6
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 60 °C
HPLC ≥97.5 %
Specific optical rotation +200° to +220° (20 °C, 589 nm) (c=3, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR