2,3-Dihydro-1H-inden-2-amine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminoindan hydrochloride, 2-Indanylamine hydrochloride

10386-0010 10386-0050 10386-0250
TS10386-0010EA 40 USD
TS10386-0010 TS10386-0050 TS10386-0250
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
2,3-Dihydro-1H-inden-2-amine hydrochloride
Formula: C₉H₁₂ClN
Melting Pt: 246…247 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012549
CAS Number: 2338-18-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR