6-Aminocaproic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

10330-0025 10330-0100 10330-1000 10330-5000
TS10330-0025EA 485 USD
TS10330-0025 TS10330-0100 TS10330-1000 TS10330-5000
6-Aminocaproic acid 99+%
6-Aminocaproic acid
Formula: C₆H₁₃NO₂
Melting Pt: 204…209 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
EINECS: 200-469-3
Merck Index: 13,00431

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 204 to 209 °C (Dec)
Solubility (13g/100ml H2O) Clear colorless solution
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR