N-Acetylglycine(Ac-Gly-OH) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid

10257-1000 10257-5000
TS10257-1000EA 37 USD
TS10257-1000 TS10257-5000
N-Acetylglycine(Ac-Gly-OH) 99%
N-Acetylglycine (Ac-Gly-OH)
Formula: C₄H₇NO₃
Melting Pt: 207…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00004275
CAS Number: 543-24-8
EINECS: 208-839-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Titration with NaOH ≥98.5 % (On dry substance)
Impurity ≤0.25 % Glycine
Sulfate (SO4) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR