Acetone cyanohydrin 99% stabilised

Supplier: Thermo Scientific Chemicals
Danger

10238-0050 10238-0250
TS10238-0050EA 56 USD
TS10238-0050 TS10238-0250
Acetone cyanohydrin 99% stabilised
Acetone cyanohydrin
Formula: C₄H₇NO
MW: 85.11
Boiling Pt: 82 °C (23 mmHg)
Melting Pt: –19 °C
Density: 0.932 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00004455
CAS Number: 75-86-5
EINECS: 200-909-4
UN: 1541
ADR: 6.1 I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20 °C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 0.3% H3PO4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR