Alcohol SDA 3C 200 Proof

Supplier: TILLEY CHEMICAL CO., INC
Danger

DNA072
TLDNA072EA 1546.69 USD
TLDNA072
Alcohol SDA 3C 200 Proof
Alcohol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR