VENOJECT® Multi-Sample Luer Adapter, Terumo
Supplier: Terumo
MN*2000T
TEMN2000CS 1232.38 USD
TEMN2000
VENOJECT® Multi-Sample Luer Adapter, Terumo
Clinical Diagnostic Supplies Clinical Instruments
Sterile, disposable adapter fits into a standard infusion set transforming it into a winged blood collection set
This allows for multiple draws into evacuated tubes. Accepts standard tube holders.