Zinc chloride ∼25% ∼2 mol/L in MeTHF (2-methyltetrahydrofuran)

Supplier: TCI America

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

Z0020-100ML
TCZ0020-100MLEA 134 USD
TCZ0020-100ML
Zinc chloride ∼25% ∼2 mol/L in MeTHF (2-methyltetrahydrofuran)
Zinc chloride

CAS Number: 7646-85-7
MDL Number: MFCD00011295
Molecular Formula: Cl2Zn
Molecular Weight: 136.28
Form: Clear Liquid
Color: Colorless

Formula: ZnCl₂
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2331
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3929e80d1118ec3a69d8caea9002b8380572e0e&vid=de30f7886a6e513bbfad24a4bb99c1ae71157d2e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR