Zinc chloride ∼7% ∼0.5 mol/L in THF

Supplier: TCI America

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

Z0019-100ML
TCZ0019-100MLEA 111 USD
TCZ0019-100ML
Zinc chloride ∼7% ∼0.5 mol/L in THF
Zinc chloride

CAS Number: 7646-85-7
MDL Number: MFCD00011295
Molecular Formula: Cl2Zn
Molecular Weight: 136.28
Form: Clear Liquid
Color: Colorless

Formula: ZnCl₂
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2331
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=643c61c11b737363381823b778ef3b50479f97d1&vid=4fb48c28cadaa8e0e0828f3f91fb3b131142058c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR