Zinc bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Zinc (II) bromide

Z0013-25G Z0013-300G
TCZ0013-25GEA 52 USD
TCZ0013-25G TCZ0013-300G
Zinc bromide ≥98.0% (by titrimetric analysis)
Zinc bromide

CAS Number: 7699-45-8
MDL Number: MFCD00011294
Molecular Formula: Br2Zn
Molecular Weight: 225.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 697
Melting point (°C): 394

Formula: Br₂Zn
Boiling Pt: 697 °C
Melting Pt: 394 °C
MDL Number: MFCD00011294
CAS Number: 7699-45-8
UN: 3260
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf82e53e0b33c6e34dae9354da3ffea74cbb1287&vid=9f54803a93bbcc85a8b9b8523cedc6a163b7e2e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR