p-Xylene ≥99.0%

Supplier: TCI America
Danger

X0044-250ML
TCX0044-250MLEA 108.1 USD
TCX0044-250ML
p-Xylene ≥99.0%
p-Xylene

[Tag Closed Flash Point Check Fluid] CAS Number: 106-42-3 MDL Number: MFCD00008556 Molecular Formula: C8H10 Molecular Weight: 106.17 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 139 Flash Point (°C): 27 Freezing point (°C): 13 Specific Gravity (20/20): 0.86

Formula: C₆H₄(CH₃)₂
Boiling Pt: 139 °C
Melting Pt: 13 °C
Density: 0.86 (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c8f5e656a3aa09650e758bcc5f79878665bbbe4&vid=36fe2c61fa128eafaabc3717414b142e92cea205&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR