Xylitol ≥98.0%
Supplier: TCI America
Synonyms:
(2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
X0018-25G
X0018-500G
TCX0018-025GEA
17.11
USD
TCX0018-025G
TCX0018-500G
Xylitol ≥98.0%
Xylitol
CAS Number: 87-99-0
MDL Number: MFCD00064292
Molecular Formula: C5H12O5
Molecular Weight: 152.15
Purity/Analysis Method:
Formula:
C₅H₁₂O₅ Boiling Pt: 495 °C (1013 hPa) Melting Pt: 92 °C Density: 1.52 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064292 CAS Number: 87-99-0 EINECS: 201-788-0 Merck Index: 13,10140 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...