Wedelolactone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1,8,9-Trihydroxy-3-methoxy-6H-benzofuro[3,2-c]chromen-6-one, 7-Methoxy-5,11,12-trihydroxy-coumestan

W0015-10MG W0015-1MG
TCW0015-10MGEA 478 USD
TCW0015-10MG TCW0015-1MG
Wedelolactone ≥98.0% (by HPLC)
Wedelolactone
Formula: C₁₆H₁₀O₇
Melting Pt: 315 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07778564
CAS Number: 524-12-9

Order Now

Specification Test Results

Appearance White to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR