Valacyclovir hydrochloride hydrate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride

V0111-100MG V0111-1G
TCV0111-100MGEA 31 USD
TCV0111-100MG TCV0111-1G
Valacyclovir hydrochloride hydrate ≥97.0% (by HPLC, total nitrogen)
Valacyclovir hydrochloride hydrate

CAS Number: 124832-27-5
MDL Number: MFCD01861507
Molecular Formula: C13H20N6O4
Molecular Weight: 360.80
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Color: White
Lambda max.: 253 nm (H2O)


Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs, Prodrug Ingredients
Other Product Names / Synonyms: L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride

Formula: C₁₃H₂₀N₆O₄
MW: 360.80 g/mol
MDL Number: MFCD01861507
CAS Number: 124832-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a26360ac4a53f9b2c56059ecfcc4b95911671f1&vid=360d27822df19258f049762735aa3020e3444184&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR