3,4-Dimethoxybenzyl alcohol ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol

V0020-25ML
TCV0020-025MLEA 50.88 USD
TCV0020-025ML
3,4-Dimethoxybenzyl alcohol ≥98.0% for synthesis
3,4-Dimethoxybenzyl alcohol

CAS Number: 93-03-8 MDL Number: MFCD00004638 Molecular Formula: C9H12O3 Molecular Weight: 168.19 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 173 Flash Point (°C): 113 Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: 3,4-Dimethoxybenzyl Alcohol

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 173 °C
Density: 1.18 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f938838a1cc60bab4e0602dddf7765b2105563ab&vid=f938838a1cc60bab4e0602dddf7765b2105563ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR