Vinylene carbonate ≥97.0% stabilized
Supplier: TCI America
(stabilized with BHT)
CAS Number: 872-36-6
MDL Number: MFCD00005380
Molecular Formula: C3H2O3
Molecular Weight: 86.05
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction), Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,3-Dioxol-2-one
Formula:
C₃H₂O₃ Boiling Pt: 60 °C Flash Pt: 73 °C |
MDL Number:
MFCD00005380 CAS Number: 872-36-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...