1-[3-(Triethoxysilyl)propyl]urea 40 - 52% in methanol

Supplier: TCI America

Synonyms: Triethoxy-3-ureidopropylsilane, 3-Ureidopropyltriethoxysilane

U0048-25ML
TCU0048-025MLEA 88.9 USD
TCU0048-025ML
1-[3-(Triethoxysilyl)propyl]urea 40 - 52% in methanol
1-[3-(Triethoxysilyl)propyl]urea

CAS Number: 23779-32-0
MDL Number: MFCD00047877
Molecular Formula: C10H24N2O4Si
Molecular Weight: 264.40
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 14


Other Product Names / Synonyms: Triethoxy-3-ureidopropylsilane, 3-Ureidopropyltriethoxysilane

Formula: C₁₀H₂₄N₂O₄Si
MW: 264.40 g/mol
Flash Pt: 14 °C
MDL Number: MFCD00047877
CAS Number: 23779-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=defb64e0aa7f6e25b83c087e44949e07e7e9a11c&vid=7d4ddede8ad5f1b260a4fce8f0a06091f50657eb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR