Sodium 1-undecanesulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
U0031-25G
TCU0031-025GEA
193.5
USD
TCU0031-025G
Sodium 1-undecanesulfonate ≥98.0% (by titrimetric analysis)
Sodium 1-undecanesulphonate
CAS Number: 5838-34-6
MDL Number: MFCD00053627
Molecular Formula: C11H24O3S
Molecular Weight: 258.35
Purity/Analysis Method:
Other Product Names / Synonyms: 1-Undecanesulfonic Acid Sodium Salt
Formula:
C₁₂H₂₅NaO₃S MW: 258.36 g/mol |
MDL Number:
MFCD00053627 CAS Number: 5838-34-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...