3,3',5,5'-Tetramethylbenzidine (TMB) solution for ELISA, ready-to-use

Supplier: TCI America

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

T3854-100ML
TCT3854-100MLEA 69 USD
TCT3854-100ML
3,3',5,5'-Tetramethylbenzidine (TMB) solution for ELISA, ready-to-use
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now

Specification Test Results

Appearance Colourless to Almost Colourlessclear liquid
Peroxidase detection test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR