Trihexyl(tetradecyl)phosphonium chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

T3623-25G T3623-5G
TCT3623-25GEA 146 USD
TCT3623-25G TCT3623-5G
Trihexyl(tetradecyl)phosphonium chloride ≥97.0% (by titrimetric analysis)
Trihexyl(tetradecyl)phosphonium chloride
Formula: C₃₂H₆₈ClP
MDL Number: MFCD04112746
CAS Number: 258864-54-9
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR