6-Quinolineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T3507-1G
TCT3507-1GEA 49 USD
TCT3507-1G
6-Quinolineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
6-Quinolineboronic acid pinacol ester
Formula: C₁₅H₁₈BNO₂
MDL Number: MFCD06411327
CAS Number: 406463-06-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR