Tetrahydrofuran-[D₈]

Supplier: TCI America
Danger

Synonyms: Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8

T3502-10ML
TCT3502-10MLEA 516 USD
TCT3502-10ML
Tetrahydrofuran-[D₈]
Tetrahydrofuran-[D8]
Formula: C₄D₈O
MW: 80.04 g/mol
Boiling Pt: 65 °C
Melting Pt: –106 °C
Flash Pt: −17 °C
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless clear liquid
Purity (GC) min. 99.0 %
Degree of deuteration min. 99.5 atom%D

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR