Tetrahexylammonium hydrogen sulfate ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

T3495-25G T3495-5G
TCT3495-25GEA 292 USD
TCT3495-25G TCT3495-5G
Tetrahexylammonium hydrogen sulfate ≥98.0% (by total nitrogen and titration analysis)
Tetrahexylammonium hydrogen sulfate
Formula: C₂₄H₅₃NO₄S
Melting Pt: 98°C
MDL Number: MFCD00037675
CAS Number: 32503-34-7
EINECS: 251-069-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR