Trimethylphosphine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Methyl phosphine

T3489-5G
TCT3489-5GEA 294 USD
TCT3489-5G
Trimethylphosphine ≥98.0% (by GC)
Trimethylphosphine
Formula: C₃H₉P
Boiling Pt: 40 °C
Melting Pt: −86 °C
Flash Pt: −30 °C
MDL Number: MFCD00008510
CAS Number: 594-09-2
EINECS: 209-823-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR