4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T3399-1G T3399-200MG
TCT3399-1GEA 311 USD
TCT3399-1G TCT3399-200MG
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Formula: C₁₇H₂₀BNO₂
MDL Number: MFCD20923942
CAS Number: 1009033-83-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR