3,4,5-Trifluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T3376-1G T3376-5G
TCT3376-1GEA 34 USD
TCT3376-1G TCT3376-5G
3,4,5-Trifluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,4,5-Trifluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₄BF₃O₂
MDL Number: MFCD05663885
CAS Number: 827614-70-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR